Short Wave Fluorescent Minerals Set
Supplier: Geoscience Industries
1560 + PKG
470151-146PK 72 USD
470151-146
Short Wave Fluorescent Minerals Set
Specimens Mineral Specimens Mineral Collections
Demonstrate the vivid contrasting colors of fifteen specimens when illuminated under shortwave ultraviolet light. Some specimens are also long wave reactive. Specimen average size: 1" x 1". Comes in compartmented box with key sheet.
  • Samples display a wide variety of fluoresence
  • Includes a sample key