Mineral ID Lab with Sort Guide
Supplier: Geoscience Industries
1761-100
470151-136EA 126.25 USD
470151-136
Mineral ID Lab with Sort Guide
Specimens Mineral Specimens Mineral Collections
Teach your students how to detect various minerals by their hardness, luster, streak and color.
Contains 20 minerals, streak plate, iron nail, magnifier and magnet. Also includes Sort Guides to help you accurately identify the sample.