Introduction to pH Measurement Lab Activity
Supplier: Lab-Aids
80
470150-612KT 97.85 USD
470150-612
Introduction to pH Measurement Lab Activity
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
Highly Graphic Introduction to the Properties of Acids and Bases
  • Observe the effects of acids and bases on indicators
  • Measure the important function of respiration
  • Requires one 50 minute class period to complete
This series of simple, highly visual lab activities introduce students to the effects of acids and bases on litmus indicators, indicator solutions, and natural indicators. The activities begin by discussing the differences between acids and bases. It then moves to a discussion on the use of pH test paper and the interpretation of their results. Students are also introduced to the concept that acid and base discussions are not limited to chemistry but also extend to applications in biology and the study of the human body. The lab activity uses LAB-AIDS® Chemplates® to study side by side, the reaction of Hydrochloric Acid and Sodium Hydroxide when mixed with other liquids and on pH paper.
Ordering information: Kit includes Chemplates® w/spatula, Hydrochloric acid, Bromothymol blue, Phenol red, Sodium hydroxide, Phenolphthalein, Blue litmus paper, Red litmus paper, and Universal indicator paper.