Jiffy® Pots
Supplier: PLANTATION PRODUCTS, INC.
JP310
470150-548CS 397.15 USD
470150-548
Jiffy® Pots
Trays/Dishes/Pots Pots
Plant roots can grow with ease through these pot walls, allowing you to replant without removing the pot. Because transfer is avoided, the plant may develop more quickly than usual.