Perfect Ink® Fingerprint Pad
Supplier: Tri-Tech
PAD-PI-5
470150-526EA 111.2 USD
470150-526
Perfect Ink® Fingerprint Pad
Educational Materials Forensics Educational Materials Fingerprint Analysis Learning Activities
Long lasting and durable ink pad.
  • Approximately 500 prints from a single pad
  • Ceramic pad
  • Easy cleanup
Perfect Ink® Fingerprint Pads contain a ceramic pad which is impregnated with a fast-drying, non-smearing, permanent black semi-inkless ink. The ceramic pad will not deteriorate with use and the non-toxic ink can be easily removed by rubbing the fingers together or wiping with a dry tissue.