Ward's® Petrographic Microscope Slides
Supplier: VWR International
2746-602765
470150-480CS 722 USD
470150-480
Ward's® Petrographic Microscope Slides
Slides Microscope Slides
Manufactured to Precise Specifications
Each noncorrosive glass slide, uniform in thickness, length, and width, measures 1" x 2", 27 x 46 mm.
Edges are beveled, ground, and polished.
Thickness: 1.2 mm.