Coliform Test Kit
Supplier: Ward's Science
1S5031
470150-462EA 66 USD
470150-462
Coliform Test Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Less confusing than other test kits that require students to distinguish different bacterial colonies on a Petri dish, this test features a distinct color change that will tell with certainty if fecal coliforms are present.
  • Students Determine the Presence or Absence of Harmful Bacteria
  • Simple to Use and Interpret
  • Instructions Included
  • Material for 25 Tests
  • Qualitative Coliform Test Yields Results within 24 Hours
The marked color change determined by E. coli presence occurs overnight in a lactose broth.
It includes materials for 25 tests and instructions.