Protein Folding Study Activity Kit
Supplier: GENO TECHNOLOGY SE
BE-411
470149-920KT 228.29 USD
470149-920
Protein Folding Study Activity Kit
Protein Extraction Protein Extraction Kits
Demonstrate Principles of Protein Denaturation and Refolding
Protein unfolding and folding, a reversible process, is analyzed using a light emitting protein specifically prepared for this study and a protein electrophoresis technique. The kit allows students to see when a light emitting protein is denatured (unfolded) as it loses its property to emit light. Materials supplied for six groups of up to five students each.