Antigen Antibody Interaction Activity Kit
Supplier: G-Biosciences
BE-501
470149-918KT 228.29 USD
470149-918
Antigen Antibody Interaction Activity Kit
Educational Materials Biology Educational Materials Cell Biology Learning Activities
Discover Properties and Processes of Antibodies
Study hands-on lab applications for antigen-antibody interaction concepts including Ouchterlony Double Diffusion, Radial Immunodiffusion, immunoelectrophoresis, and immunoprecipitation. Students will discover specific properties and processes of antigens and antibodies, including diffusion, interaction, and complex formation. Materials supplied for six student groups for up to five students each.