Size Exclusion Chromatography Activity Kit
Supplier: G-Biosciences
BE-414
470149-898KT 183.19 USD
470149-898
Size Exclusion Chromatography Activity Kit
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
Demonstrate Principles and Factors of Gel Filtration
This hands-on chromatography lab activity teaches gel filtration or size exclusion chromatography and the use of this method in purification of biological samples. Materials supplied for six student groups of up to five students each.