Make Your Own Watershed Kit
Supplier: JT & A, INC.
71005K-B
470149-796CS 652.75 USD
470149-796
Make Your Own Watershed Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Modeled after the award-winning EnviroScape models, this kit allows your students to create their own city and learn about water pollution at the same time.
  • A great hands-on way to explore the environment
Instructional activities highlight how water pollution occurs and how to prevent it — such as adding cocoa for soil, drink mixes for chemicals, and making it rain. You can even make manure for use as a pollutant on the model – and experiment with techniques (best management practices) for preventing water pollution.
The kit includes plaster cast forms, three paint brushes, non-toxic acrylic paint, three felt strips, bridges, instructions, and activities.