Permanent Ceramic Block Magnet
Supplier: Master Magnetics
CB60CMAG
470149-774EA 1.05 USD
470149-774
Permanent Ceramic Block Magnet
Magnets
A permanent ceramic block magnet with maximum magnetic efficiency, high inductance, and a minimum stray magnetic field.
The magnet is magnetized through the thickness to have complete freedom from pole orientation. Size: 50×25×10 mm.
WARNING: KEEP AWAY FROM ALL CHILDREN! This product contains (a) small magnet(s). Swallowed magnets can stick together across intestines causing serious infection and death. Seek immediate medical attention if magnet(s) are swallowed or inhaled.