Equal Volume Density Set, Rectangular
Supplier: Wards
PH0111
470149-614KT 29.45 USD
470149-614
Equal Volume Density Set, Rectangular
Educational Materials Physics Educational Materials Mechanics Learning Activities
Rectangular density set.
  • Set of 4 equal volume rectangular prisms
  • Includes: Aluminum, Copper, Steel and Brass
  • Dimensions: 1.25×1.25×5 cm (0.5×0.5×2")
  • Machine tooled to the most exacting tolerances in the industry
This 4 piece set includes aluminum, brass, copper and steel specimens of equal volume designed for successful density experiments and specific gravity measurement.