96-Place Microcentrifuge Tube Rack
Supplier: Ward's Science
241014
470149-570CS 162.3 USD
470149-570
96-Place Microcentrifuge Tube Rack
Racks Tube Racks
High-volume rack
This rack is designed with wide spacing to allow storage of ninety six 1.5 or 2 ml tubes.The bright racks are designed for easy storage of microcentrifuge tubes. There are 11 mm diameter holes on one side and 9 mm on the other.
Ordering information: Color may vary from image.