Prisms
Supplier: VWR International
P30273
470149-510CS 500 USD
470149-510
Prisms
Educational Materials Physics Educational Materials Optics Learning Activities
Clear, Short Prisms for Index of Refraction and Dispersion
Equilateral prisms are clear with long sides for refractive index measurements, trapezoidal prisms have a frosted base to show ray paths inside the prism.