Density Identification Set
Supplier: Shiv Dial Sud & Sons
P10043A
470149-492CS 1744.6 USD
470149-492
Density Identification Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
The Density Identification Set is ideal for an investigation lab.
  • Twelve Different Specimens to Test
  • Aluminum Cylinders Included
  • Specimens Delivered in Wood Storage Block
  • Linseed Oil Coating Protects Block from Wet Specimens
  • Ideal for Investigation Labs
This density-cylinder set consists of twelve half-inch dia. specimens of varying lengths and materials.
The specimens are supplied in an attractive wooden storage block. The block is protected from the wet specimens by a linseed oil coating.
Ordering information: The set includes cylinders of aluminum, brass, copper, Pyrex glass, hard rubber, Delrin, PVC, acrylic, Teflon, and Nylon plastics, and poplar and oak wood.
Packaging: One set includes 12 specimens.