Ten Specimen Density Set
Supplier: Shiv Dial Sud & Sons
P10040
470149-490CS 2288.82 USD
470149-490
Ten Specimen Density Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
This set features a handy, economical selection of density cubes.
  • Economical
  • Handy
  • Cubes Measure 1" Each
  • Materials Include Pine, Poplar, Plastic, and More
  • Ten Density Cubes per Set
The cubes measure 1" each.
They're made of acrylic plastic, aluminum, brass, copper, nylon, oak, pine, poplar, PVC, and steel.