Acrylic Ray Optics Set
Supplier: VWR International
P30278
470149-474CS 613.1 USD
470149-474
Acrylic Ray Optics Set
Educational Materials Physics Educational Materials Optics Learning Activities
A Good Optics Set for Demonstrations or First Experiments
  • Made of polished acrylic
  • Provided in a convenient storage box
Set consists of: rectangle (75mm slides), double convex lens and two double concave lenses with a focal length of about 10cm, equilateral prism, and semi-circular block. 15mm thick.