Squibb Urinometer and Jar
Supplier: Shiv Dial Sud & Sons
P11098-B
470149-468CS 188.72 USD
470149-468
Squibb Urinometer and Jar
Clinical Diagnostic Supplies Clinical Instruments
For Clean and Accurate Results
For highly accurate samples when only minimal volumes are required, use this simple, stain-free apparatus. Made of high-quality clear glass, this 50 mL urinometer has a wide bottom to prevent tipping and a non-stick float. It includes instructions. Range: 1.000–1.060 Sp. Gr in 0.002 graduations.