Stainless Steel Spatula
Supplier: Shiv Dial Sud & Sons
CH12229A/1
470149-442CS 420.85 USD
470149-442
Stainless Steel Spatula
Spatulas
Double Use Utensil.
  • Stainless steel
  • 9" length
  • One end spoon shaped - 13 mm wide
  • One end flat - 50×8 mm wide
Easily remove loose chemicals from small bottles with minimal spillage. It also can be used as a stirrer or for breaking up clumped material.