Vacuum Plate and Pad
Supplier: Shiv Dial Sud & Sons
PH36018A/3 PH36018A/1
470149-426CS 1378.41 USD
470149-426 470149-428
Vacuum Plate and Pad
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
This vacuum plate and pad are the perfect tools for vacuum experiments.
  • No Messy Vacuum Wax Needed
  • Air-Tight Contact
  • Ideal with Bell Jars and Vacuum Pumps
  • Center Hole: 50 mm
  • Safety Screw Plug
They work with bell jars and vacuum pumps, and no messy vacuum wax is needed. The handy plate features a 50 mm center hole, safety screw plug, and stopcock with hose nipple for vacuum tubing of 1/4" ID.
The top plate measures 11" in diameter. It's made of cast iron.