Centripetal Force Kit
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SV583
470149-398CS 190.6 USD
470149-398
Centripetal Force Kit
Educational Materials Physics Educational Materials Mechanics Learning Activities
The Centripetal Force Kit is inexpensive and effective.
  • Designed for PSSC
  • Plastic Tube
  • Two Rubber Stoppers
  • Steel Washers
  • Instructions Included
This kit was originally designed for PSSC, and it provides excellent data with centripetal force.
A plastic tube works as the pivot, two rubber stoppers are the whirling mass, steel washers are the counterforce, and the nylon cord ties the stoppers to the washers.
Ordering information: This kit includes complete instructions.