Multimeter
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SE067A
470149-374CS 218.05 USD
470149-374
Multimeter
Electrical Meters Digital Multimeters
Multiple features and a large LCD display make this meter ideal for beginning electronics courses.
  • Transistor and Diode Testing
  • Overload Protection
  • LCD Display
  • Battery and Test Leads
  • Ideal for Beginning Electronics Courses
The meter includes volts, ohms, resistance, and current testing plus transistor and diode testing.
The meter features overload protection and includes battery and test leads. Instructions are included.