VWR® Petri Dish Sets (Tops and Bottoms)
Supplier: VWR International
470149-320 470149-322
470149-320CS 680 USD
470149-320 470149-322
VWR® Petri Dish Sets (Tops and Bottoms)
Culture Dishes Petri Dishes
Our high-quality borosilicate Petri dishes meet ASTM standards and are a great value.
  • High-quality fabrication
  • Borosilicate glass
  • Meet ASTM standards
Petri dish sets include covers.