VWR® Volumetric Flasks with Glass Stoppers, Class B
Supplier: VWR International
470149-302
470149-302CS 805 USD
470149-302
VWR® Volumetric Flasks with Glass Stoppers, Class B
Flasks Volumetric Flasks
Flasks feature a permanent graduation line and large marking area, and necks are tooled to fit the included glass stoppers.
  • Borosilicate glass fabrication
  • Large marking area
  • Glass stopper included
With tolerances twice those specified for Class A glassware, these Class B borosilicate volumetric flasks are a great value.