VWR® Wide-Mouth Erlenmeyer Flasks
Supplier: VWR International
470149-296 470149-298 470149-300 470149-318
470149-296CS 317.83 USD
470149-296 470149-298 470149-300 470149-318
VWR® Wide-Mouth Erlenmeyer Flasks
Flasks Erlenmeyer Flasks
High Quality Borosilicate Glass.
  • Meet ASTM standards
  • Uniform wall thickness
  • White enamel metric scale, and marking spot
Can be heated and used with acids, salt solutions, and organic solvents.
Ordering information: Rubber stoppers are not included.