VWR® Florence Boiling Flasks
Supplier: VWR International
470149-284 470149-286 470149-288 470149-290
470149-284CS 151.03 USD
470149-284 470149-286 470149-288 470149-290
VWR® Florence Boiling Flasks
Flasks Round Bottom Flasks
High Quality Borosilicate Glass
  • Meet ASTM standards
  • Round body and long neck are ideal for boiling liquids without excessively rapid evaporation
Ordering information: Rubber stoppers are not included.