VWR® Culture Tubes
Supplier: VWR International
470149-242 470149-244 470149-246 470149-248 470149-250 470149-252 470149-254 470149-256
470149-242CS 652.81 USD
470149-242 470149-244 470149-246 470149-248 470149-250 470149-252 470149-254 470149-256
VWR® Culture Tubes
Tubes Culture Tubes
These culture tubes are made of high-quality borosilicate glass.
  • Rimless for easier plugging
Each tube is alkali-free and inert, so it will not affect the pH of media.
Certifications: ASTM standards.
Accessories information: Caps sold separately.