Insect Pinning Block
Supplier: VWR International
INSPN-BLK
470149-222CS 107 USD
470149-222
Insect Pinning Block
Specimen Mounting Supplies
Traditional, 3-step wood pinning block has pin holes set at 23, 12 and 7 mm. Size: 1" x 3".