Hand Stroboscope
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SH591A
470149-132CS 181.65 USD
470149-132
Hand Stroboscope
Educational Materials Physics Educational Materials Mechanics Learning Activities
This hand stroboscope is a cardboard disk with a diameter of 24 cm and 12 slots.
The center, rotating, ball bearing mount has a holding disk that students grip while they spin the disk with the finger hole.