Graphite Coated Spheres
Supplier: VWR International
PH45244A
470149-122CS 318.9 USD
470149-122
Graphite Coated Spheres
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Used to Detect Electrostatic Charge
Coated pith balls and spheres have a light conductive coating used in charge transfer experiments.