Fundamentals of Physics Projectile Launcher

Supplier: Wards
PRLN01 BLP-FPL BST-FPL
470014-468EA 188.16 USD
470014-468 470149-100 470149-106
Fundamentals of Physics Projectile Launcher
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
This unique metal launcher has multiple settings, so launch speed and angles may vary.

  • Launcher shoots the ball at any angle from –15 to 90°
  • Sturdy Wooden Base
  • Length: 15¾"
  • Width: 11¾"
  • Height: 4"

A large, easy-to-read protractor allows any angle between 0 and 90° to be set securely. The sturdy wood base allows the photogate system to be attached at the launch end, regardless of the angle.

Marbles, instructions, and an activity guide are included. The launcher measures 15¾ x 11¾ x 4".

Caution: This product has small parts. It is not for children under three years. CHOKING HAZARD. Use eye protection.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR