VWR® Volumetric Flasks
Supplier: VWR International
470149-066 470149-104 470190-862 470191-144 470191-256 470191-264 470191-268
470149-066CS 623.07 USD
470149-066 470149-104 470190-862 470191-144 470191-256 470191-264 470191-268
VWR® Volumetric Flasks
Flasks Volumetric Flasks
A Great Value!
  • Class B borosilicate glass
  • Includes choice of glass stopper or snap cap
  • Graduation line for accurate measurements
Our new Class B borosilicate volumetric flasks feature capacity tolerances twice those specified for Class A volumetric ware.