Hero’s Engine
Supplier: VWR International
PH20505A
470149-036CS 463.3 USD
470149-036
Hero’s Engine
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Students Can Simulate the Earliest Steam Engine.
  • Includes a flask, chain, and rubber stopper
Suspended by a chain over a flame, this apparatus simulates the earliest form of steam engine (100 A.D.). The steam produced is ejected from the 125 ml flask’s side nozzles causing the flask to rotate.