High Temperature Burners
Supplier: Shiv Dial Sud & Sons
C-1490/1
470149-024CS 1136.73 USD
470149-024
High Temperature Burners
Heaters Gas Burners
These Meker type burners are for use where an extremely hot flame is required.
  • Grid diameter is 4 cm (1¹/₂")
  • Overall height is about 20 cm (8")
  • Use 9.5 mm (³/₈") tubing
The perforated monel grid has a large surface area to produce a short, intense flame. The needle valve gas regulator is easily accessible, facilitating fine adjustment.