Polarized Film Slides
Supplier: Shiv Dial Sud & Sons
P30695/3
470149-018CS 4692.28 USD
470149-018
Polarized Film Slides
Educational Materials Physics Educational Materials Optics Learning Activities
Learn how sunglasses work!
  • Pair of identical slides with polarized film
When this pair of polaroid film squares are aligned, light passes through them appearing much as it would if viewed through sunglasses. However, if one square is rotated 90 degrees, no light passes through at all! A great way to demonstrate the principles of polarization and the wave properties of light.