Acceleration Trolley
Supplier: VWR International
P10515A
470149-004CS 771.1 USD
470149-004
Acceleration Trolley
Educational Materials Physics Educational Materials Mechanics Learning Activities
Show your students how the rate of acceleration of an object depends on the angle of incline with this great hands-on activity.
  • Simply fasten to your blackboard or wall and string a cable across the classroom at any angle
Mount the trolley on the cable, add weights with holes provided for weight hangers, and use a stopwatch to measure the time it takes for the trolley to cross.
Includes: trolley with pulley, 9.1m (30 ft.) wire cable, attachment kit with hardware, and instructions.