Newton’s Color Disc
Supplier: VWR International
PH30701A
470148-974CS 454.3 USD
470148-974
Newton’s Color Disc
Educational Materials Physics Educational Materials Optics Learning Activities
What Happens When You Combine All Colors of Light?
  • Great classroom visual
  • Ideal for Elementary and Middle School levels
Easily demonstrate that all the colors of the visible spectrum can be combined to form white light with this simple device. Comprised of a pulley mounted on a metal stand, the seven colored metal disc is driven by a belt connected to a hand-driven wheel.