Leak-Proof Wickless Burner
Supplier: Shiv Dial Sud & Sons
CH10652
470148-968CS 820.75 USD
470148-968
Leak-Proof Wickless Burner
Heaters Spirit Lamps
Tip Resistant.
  • Unit holds 100 ml of fuel that will burn for about 1½ hours
  • Height: 3¼"
This tip-resistant alcohol burner uses a copper-plated torch instead of a wick to ensure even heat distribution. Even if tipped upside down, the burner will not leak. It comes with an aluminum snuffer cap.