Collision in Two Dimensions
Supplier: VWR International
P10505
470148-956CS 985.8 USD
470148-956
Collision in Two Dimensions
Educational Classroom Kits and Activities
The Collision in Two Dimensions apparatus is a rich source of data for studying conservation of momentum and conservation of kinetic energy in collisions.
  • For Studying a Variety of Scenarios
  • Inexpensive
  • Eye Screw Works a Plumb Bob
  • One Glass Eye Included
  • Apparatus Also Compares Elastic and Inelastic Collisions
It is also used to compare elastic and inelastic collisions.
A curved metal track and an adjustable target support from the core of the unit.
Ordering information: This kit includes two steel balls, a glass ball, and an eye screw, which serves as a plumb bob. It requires a C-clamp for mounting on a table, available separately.