Bottles, Polypropylene, Narrow Mouth
Supplier: VWR International
470148-782 470148-946 470149-136 470149-594 470149-596 470200-822 470201-066 470201-068
470148-782CS 250.31 USD
470148-782 470148-946 470149-136 470149-594 470149-596 470200-822 470201-066 470201-068
Bottles, Polypropylene, Narrow Mouth
Bottles Plastic Bottles
Leak-proof.
  • Round
  • Polypropylene screw cap supplied
  • May be autoclaved empty or filled