Dual Microtube Rack with Lid
Supplier: Ward's Science
241010
470148-892CS 126.4 USD
470148-892
Dual Microtube Rack with Lid
Racks Tube Racks
Space-Saving, Economical Unit
This versatile, dual-sided unit accommodates ninety-six 1.5–2.0 ml microcentrifuge tubes or 0.5 ml PCR Tubes. Polypropylene rack is autoclavable and includes handles and a lid for safe transport and storage. Colors may vary.