2 Battery Holder with Plug Jacks
Supplier: OSAW INDUSTRIAL PRODUCTS SE
S0598
470148-864CS 258.6 USD
470148-864
2 Battery Holder with Plug Jacks
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A simple tool for communicating complex workings of electricity, this plastic battery holder is fitted with three banana plug jacks.
  • Illustrate DC, series, open, and closed circuits
  • Easy switch between selections of DC, 1.5 and 3 V power for your experiments
The holder fits two D-cell batteries and can be used to demonstrate circuits, Ohm's law, and the transformation of energy. Batteries not included.