Clamp Pulley
Supplier: Shiv Dial Sud & Sons
P10324
470148-858CS 482.8 USD
470148-858
Clamp Pulley
Clamps General Purpose Clamps
This free-running pulley features a rugged plastic sheave and deep V grooves.
  • Pulley Clamps to Surfaces up to One-Inch Thick
  • Rugged Plastic Sheave
  • Deep V Grooves
  • Free Running
It can clamp to surfaces up to one-inch thick.
The sheave is two inches in diameter.