VWR® Double-Scale Cylinders
Supplier: VWR International
470148-852
470148-852CS 280.3 USD
470148-852
VWR® Double-Scale Cylinders
Cylinders Measuring Cylinders
Our high-quality borosilicate cylinders are sturdy and easy to use.
  • Uniform Wall Thickness
  • White Enamel Fabrication
  • Borosilicate Glass Construction
  • Dual-Graduated Metric Scale Numbered in Both Directions
  • Molded Base
They can be used with acids, salt solutions, and organic solvents.
The dual-graduated metric scale is numbered in both directions.