Adjustable Micro Bunsen Burners
Supplier: Shiv Dial Sud & Sons
C-1486
470148-840CS 521.29 USD
470148-840
Adjustable Micro Bunsen Burners
Heaters Gas Burners
Ideal for Jobs Requiring a Small, Intense Flame
  • Nickel-plate, die-cast zinc alloy base
  • Flame stabilizer and needle valve for precise flame control
  • Gas inlet is serrated for secure connections
Gas inlet 5/16" outer dia.; mixing tube 7/16" outer dia.; 3 7/8"H