Quantitative Electroscope
Supplier: VWR International
P45259
470148-820CS 658.9 USD
470148-820
Quantitative Electroscope
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Electrostatic charges are indicated by the deflection of a black plastic straw pivoted on a pin. The polished zinc plate on top is used to demonstrate photoelectric effect and various other electrostatic demonstrations. The metal case measures 14 cm in diameter. The scale is graduated in 10° increments in two quadrants.