Differential and Sympathetic Tuning Forks
Supplier: Shiv Dial Sud & Sons
P36140
470148-808CS 334.88 USD
470148-808
Differential and Sympathetic Tuning Forks
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
These large alloy tuning forks are designed to demonstrate resonance, beat frequency, and sympathetic vibration.
  • Frequency: 256Hz
  • Moveable Mass Changes Frequency
  • Forks Demonstrate Resonance, Beat Frequency, and Sympathetic Vibration
Each fork has a frequency of 256Hz. One fork has a movable mass to change the frequency. The closer the frequency is to the other tuning fork, the slower the beat pattern.
Students can see the beat patters using a probeware microphone. Assembly is required.