21st Century Balance
Supplier: VWR International
P10854
470148-796CS 471.5 USD
470148-796
21st Century Balance
Educational Materials Physics Educational Materials Mechanics Learning Activities
This balance is the first major advancement in teaching simple machines in decades.
  • Unique Features
  • Internal Sliding Weight Mechanism
  • Instructions Included
  • Scale: 50 cm
The internal sliding weight mechanism allows users to neutralize all factors once the fulcrum and positions of force have been established.
It includes instructions and a 50 cm scale.