Density ID Set
Supplier: Shiv Dial Sud & Sons
PH10044A
470148-790CS
949.9
USD
470148-790
Density ID Set
Educational Materials
Physics Educational Materials
Mechanics Learning Activities
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...